Display Info New header:
tfpt-header-footer.tfcom-eks-na-comerce-gateway-prd.commerceprod.thermofisher.net cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@770c7432
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous enableSimpleHeaderValue:
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search All
Popular Product Areas
TaqMan Assays
Gene Expression
Microbe & ABR Detection
Copy Number Variation
Mutation Detection
miRNA
SNP Genotyping
Primary Antibodies
Secondary Antibodies
Isotype Controls
Proteins & Peptides
ELISA Kits
All Documents & Support
Certificates
SDS
Manuals & Protocols
Product FAQs
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OBSCN SIRNA Assays ›
Gene Symbol:
OBSCN
Gene Name:
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases:
ARHGEF30, UNC89
Chromosome Location:
Chr.1: 228208013 - 228381431 on Build GRCh38
UniGene ID:
Hs.650039
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s38404
Purification
HPLC
Standard
HPLC-IVR
Size
20 nmol
40 nmol
250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
40 nmol
250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price (USD)
388.00
Your Price
Online offer (USD):
374.65
374.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
Displaying...
Build 38.2 Human Chr.1: s38404
×
Created with Raphaël 2.1.2
IBA57
C1orf145
OBSCN
LOC10192
7401
TRIM11
MIR6742
TRIM17
NM_00101
0867.3
NM_00131
0327.1
NR_07315
5.1
NR_07315
4.1
XM_01154
4352.2
XM_01154
4351.2
XM_01154
4285.2
XM_01700
2412.1
XM_01700
1418.1
XM_01154
4209.2
HY081013
.1
AK022796
.1
HY203715
.1
CA425016
.1
AW875576
.1
BF439536
.1
BC027909
.1
AK056556
.1
BX335215
.2
BX335216
.2
AK024186
.1
BC098118
.1
AK128447
.1
BU683616
.1
DB163213
.1
AK097489
.1
AB046776
.1
AL080128
.1
CN302348
.1
AL832357
.1
HY192178
.1
AK301859
.1
AW770403
.1
BM563820
.1
227,700,
000
227,800,
000
227,900,
000
228,000,
000
228,100,
000
228,200,
000
228,300,
000
228,400,
000
228,500,
000
228,600,
000
228,700,
000
228,800,
000
228,900,
000
Show all RefSeq and GenBank
Back To Top
siRNA Details
Gene Symbol: OBSCN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098623.2
NP_001092093.2
45
12071
NM_001271223.2
NP_001258152.2
56
14942
NM_052843.3
NP_443075.3
45
12071
XM_005273287.4
XP_005273344.1
55
14746
XM_005273291.4
XP_005273348.1
55
14746
XM_005273298.4
XP_005273355.1
55
14746
XM_005273307.4
XP_005273364.1
55
14746
XM_006711822.3
XP_006711885.1
55
14746
XM_006711823.3
XP_006711886.1
55
14746
XM_006711827.3
XP_006711890.1
49
13161
XM_006711829.3
XP_006711892.1
48
12885
XM_011544297.2
XP_011542599.1
49
13162
XM_011544299.2
XP_011542601.1
48
12886
XM_017002443.1
XP_016857932.1
55
14746
XM_017002444.1
XP_016857933.1
55
14709
XM_017002445.1
XP_016857934.1
55
14746
XM_017002446.1
XP_016857935.1
55
14746
XM_017002447.1
XP_016857936.1
55
14746
XM_017002448.1
XP_016857937.1
55
14596
XM_017002449.1
XP_016857938.1
54
14483
XM_017002450.1
XP_016857939.1
54
14483
XM_017002451.1
XP_016857940.1
54
14483
XM_017002452.1
XP_016857941.1
54
14483
XM_017002453.1
XP_016857942.1
54
14470
XM_017002454.1
XP_016857943.1
54
14470
XM_017002455.1
XP_016857944.1
54
14470
XM_017002456.1
XP_016857945.1
54
14470
XM_017002457.1
XP_016857946.1
54
14470
XM_017002458.1
XP_016857947.1
53
14219
XM_017002459.1
XP_016857948.1
53
14219
XM_017002460.1
XP_016857949.1
53
14219
XM_017002461.1
XP_016857950.1
55
14746
XM_017002462.1
XP_016857951.1
53
14194
XM_017002463.1
XP_016857952.1
53
14173
XM_017002464.1
XP_016857953.1
55
14746
XM_017002465.1
XP_016857954.1
52
13955
XM_017002466.1
XP_016857955.1
52
13955
XM_017002467.1
XP_016857956.1
52
13955
XM_017002468.1
XP_016857957.1
51
13691
XM_017002469.1
XP_016857958.1
49
13163
XM_017002470.1
XP_016857959.1
44
11844
XM_017002471.1
XP_016857960.1
55
14745
XM_017002472.1
XP_016857961.1
55
14745
XM_017002473.1
XP_016857962.1
55
14745
GenBank mRNA
AJ002535.1
-
46
12068
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
multicellular organism development
regulation of Rho protein signal transduction
protein localization to M-band
positive regulation of apoptotic process
positive regulation of GTPase activity
sarcomere organization
regulation of small GTPase mediated signal transduction
mitophagy in response to mitochondrial depolarization
protein serine/threonine kinase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
structural constituent of muscle
ankyrin binding
titin binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:/anonymous
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-59d6677d6-9mspm:80/100.66.73.217:80.
git-commit: 1e3318a2800c8d0a8829a508293490082e5b9833
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.30.0-Offline