Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMILIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032048.2 | NP_114437.2 | 2 | 313 | ||
XM_017026038.1 | XP_016881527.1 | 2 | 300 | |||
GenBank mRNA | AF270513.1 | - | 2 | 313 | ||
AI356740.1 | - | 2 | 304 | |||
AL552384.3 | - | 2 | 83 | |||
BC032231.1 | - | 2 | 300 | |||
BC136541.1 | - | 2 | 267 | 9020531 |