Gene Symbol: DTNBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271667.1 | NP_001258596.1 | 3 | 374 | ||
NM_032122.4 | NP_115498.2 | 3 | 329 | |||
NM_183040.2 | NP_898861.1 | 3 | 329 | |||
NR_036448.1 | 4 | 468 | ||||
XM_005249447.4 | XP_005249504.1 | 4 | 330 | |||
GenBank mRNA | AF394226.1 | - | 3 | 246 | ||
AK054593.1 | - | 3 | 298 | |||
AK290718.1 | - | 3 | 246 | |||
AK310590.1 | - | 4 | 330 | |||
AL136637.2 | - | 3 | 246 | |||
AY265460.1 | - | 3 | 253 | |||
BC011912.2 | - | 3 | 244 | 4139934 | ||
BQ108775.1 | - | 3 | 311 | |||
BX394617.2 | - | 4 | 468 | |||
KU178801.1 | - | 3 | 140 |