Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032141.3 | NP_115517.1 | 2 | 122 | ||
XM_017025209.1 | XP_016880698.1 | 2 | 129 | |||
XM_017025210.1 | XP_016880699.1 | 2 | 952 | |||
XM_017025211.1 | XP_016880700.1 | 3 | 150 | |||
GenBank mRNA | AF161358.1 | - | 2 | 101 | ||
AK094552.1 | - | 111 | ||||
AK313963.1 | - | 2 | 109 | |||
AL136806.1 | - | 2 | 113 | |||
BC016313.1 | - | 3 | 136 | |||
BC040118.1 | - | 2 | 102 | 5176197 | ||
BC046116.2 | - | 2 | 124 | |||
BC062425.1 | - | 2 | 114 | |||
BC105044.1 | - | 2 | 109 | 8144047 | ||
BC105046.1 | - | 2 | 109 | 8144049 | ||
CB957325.1 | - | 2 | 116 | |||
CR533555.1 | - | 2 | 85 |