Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM188B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032222.2 | NP_115598.2 | 3 | 333 | ||
GenBank mRNA | AF538962.1 | - | 3 | 326 | ||
AK026027.1 | - | 3 | 333 | |||
AK128558.1 | - | 1 | 2517 | |||
BC098569.1 | - | 3 | 280 | 30924492 | ||
DA868176.1 | - | 3 | 333 |
Gene Symbol: INMT-FAM188B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037598.1 | 5 | 785 | |||
GenBank mRNA | AK308762.1 | - | 5 | 783 |