Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TXNDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098529.1 | NP_001091999.1 | 2 | 1932 | ||
NM_032243.5 | NP_115619.4 | 2 | 1729 | |||
XM_017026041.1 | XP_016881530.1 | 3 | 1603 | |||
GenBank mRNA | AF080095.1 | - | 2 | 1462 | ||
AK097656.1 | - | 2 | 1682 | |||
AK303045.1 | - | 4 | 1284 | |||
AL136742.1 | - | 1 | 308 | |||
BC050132.1 | - | 2 | 1517 | 5164861 | ||
EF560747.1 | - | 3 | 1563 |