Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286211.1 | NP_001273140.1 | 4 | 799 | ||
NM_001286213.1 | NP_001273142.1 | 3 | 666 | |||
NM_032256.2 | NP_115632.1 | 4 | 799 | |||
XM_011538831.2 | XP_011537133.1 | 4 | 808 | |||
XM_011538832.2 | XP_011537134.1 | 4 | 483 | |||
XM_017020022.1 | XP_016875511.1 | 4 | 390 | |||
XM_017020023.1 | XP_016875512.1 | 5 | 631 | |||
GenBank mRNA | AK057287.1 | - | 4 | 518 | ||
AL136855.1 | - | 4 | 501 | |||
BC060798.1 | - | 4 | 562 | 30348706 |