Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031727.2 | NP_001026897.1 | 5 | 952 | ||
NM_032285.2 | NP_115661.1 | 5 | 811 | |||
XM_011528355.2 | XP_011526657.1 | 5 | 887 | |||
XM_011528356.2 | XP_011526658.1 | 5 | 975 | |||
GenBank mRNA | AL834276.1 | - | 5 | 802 | ||
AY358176.1 | - | 5 | 929 | |||
BC001703.1 | - | 5 | 926 | 3501899 |