Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031727.2 | NP_001026897.1 | 6 | 1933 | ||
NM_032285.2 | NP_115661.1 | 6 | 1792 | |||
XM_011528355.2 | XP_011526657.1 | 6 | 1868 |