Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FYTTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011537.2 | NP_001011537.2 | 9 | 972 | ||
NM_032288.6 | NP_115664.2 | 8 | 1031 | |||
NR_027840.1 | 8 | 1142 | ||||
GenBank mRNA | AJ344094.1 | - | 8 | 877 | ||
AK027672.1 | - | 8 | 915 | |||
AK296283.1 | - | 7 | 733 | |||
AK302546.1 | - | 8 | 1142 | |||
AK303462.1 | - | 9 | 972 | |||
AK315019.1 | - | 8 | 880 | |||
AK316308.1 | - | 8 | 772 | |||
AL136558.1 | - | 8 | 884 | |||
BC035006.2 | - | 8 | 1034 | 5267824 | ||
BC039734.1 | - | 8 | 941 | 6042926 | ||
CR533502.1 | - | 8 | 812 |