Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032301.2 | NP_115677.2 | 8 | 1263 | ||
XM_005260096.4 | XP_005260153.1 | 7 | 1382 | |||
GenBank mRNA | AK123037.1 | - | 8 | 1262 | ||
AK302744.1 | - | 6 | 988 | |||
AY736034.1 | - | 8 | 1263 | |||
BC004290.1 | - | 8 | 1240 |