Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032314.3 | NP_115690.3 | 4 | 644 | ||
XM_006719639.2 | XP_006719702.1 | 5 | 744 | |||
GenBank mRNA | AK222610.1 | - | 4 | 634 | ||
AK225242.1 | - | 4 | 634 | |||
AK293656.1 | - | 4 | 665 | |||
AK298214.1 | - | 3 | 422 | |||
AK315681.1 | - | 4 | 635 | |||
BC004916.2 | - | 4 | 620 | |||
BC107874.1 | - | 4 | 638 | 6452052 | ||
BX647562.1 | - | 4 | 633 | |||
EU700459.1 | - | 4 | 638 |