Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM213A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243778.1 | NP_001230707.1 | 2 | 215 | ||
NM_001243779.1 | NP_001230708.1 | 2 | 285 | |||
NM_001243780.1 | NP_001230709.1 | 2 | 781 | |||
NM_001243781.1 | NP_001230710.1 | 2 | 298 | |||
NM_001243782.1 | NP_001230711.1 | 2 | 198 | |||
NM_032333.4 | NP_115709.3 | 2 | 315 | |||
XM_011540265.2 | XP_011538567.1 | 2 | 618 | |||
XM_011540266.2 | XP_011538568.1 | 2 | 1021 | |||
XM_017016775.1 | XP_016872264.1 | 2 | 302 | |||
GenBank mRNA | AK027858.1 | - | 1 | 2403 | ||
AK074643.1 | - | 2 | 260 | |||
AK075447.1 | - | 2 | 290 | |||
AK225387.1 | - | 2 | 189 | |||
AK315440.1 | - | 2 | 209 | |||
AY359059.1 | - | 2 | 208 | |||
BC005871.2 | - | 2 | 177 | 3010078 | ||
BE742158.1 | - | 2 | 285 | |||
CN396006.1 | - | 2 | 167 | |||
DA136257.1 | - | 2 | 199 | |||
DB494833.1 | - | 2 | 315 |