Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032340.3 | NP_115716.1 | 2 | 235 | ||
GenBank mRNA | AF086234.1 | - | 2 | 157 | ||
AK311835.1 | - | 2 | 184 | |||
AY568085.1 | - | 2 | 211 | |||
BC006007.1 | - | 2 | 211 | 4284544 | ||
BI604191.1 | - | 2 | 185 | |||
DA123098.1 | - | 2 | 235 | |||
EF139858.1 | - | 4 | 485 |