Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT16L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193452.1 | NP_001180381.1 | 3 | 1284 | ||
NM_032349.3 | NP_115725.1 | 3 | 1214 | |||
XM_005255633.4 | XP_005255690.1 | 2 | 4286 | |||
GenBank mRNA | AK092642.1 | - | 2 | 1908 | ||
BC006223.2 | - | 3 | 1192 | 3944355 |