Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032382.4 | NP_115758.3 | 4 | 1588 | ||
GenBank mRNA | AK025968.1 | - | 2 | 710 | ||
AK056344.1 | - | 4 | 1576 | |||
AK225391.1 | - | 2 | 710 | |||
AK302605.1 | - | 4 | 1601 | |||
BC017492.1 | - | 2 | 392 | |||
BC121022.1 | - | 4 | 1548 | 40117369 | ||
BC121023.1 | - | 4 | 1548 | 40117370 |