Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303542.2 | NP_001290471.2 | 4 | 744 | ||
NM_032932.5 | NP_116321.2 | 6 | 1037 | |||
GenBank mRNA | AB058724.1 | - | 6 | 726 | ||
AJ314646.2 | - | 6 | 726 | |||
AK074060.1 | - | 2 | 505 | |||
AK097424.1 | - | 4 | 687 | |||
AY169244.1 | - | 6 | 1037 | |||
BC093914.1 | - | 6 | 840 | 7939759 | ||
BC101517.1 | - | 6 | 840 | 8069023 | ||
BI461640.1 | - | 4 | 751 |