Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318099.1 | NP_001305028.1 | 2 | 421 | ||
NM_001318100.1 | NP_001305029.1 | 2 | 498 | |||
NM_001318101.1 | NP_001305030.1 | 1 | 359 | |||
NM_032429.3 | NP_115805.1 | 2 | 528 | |||
XM_005270222.4 | XP_005270279.1 | 2 | 760 | |||
XM_017016780.1 | XP_016872269.1 | 1 | 2889 | |||
XM_017016781.1 | XP_016872270.1 | 2 | 529 | |||
GenBank mRNA | AB058716.1 | - | 1 | 3380 | ||
AK097997.1 | - | 1 | 151 | |||
AK298208.1 | - | 2 | 386 | |||
AY029201.1 | - | 1 | 242 | |||
BC006212.2 | - | 2 | 386 | 3161855 | ||
BC058938.1 | - | 2 | 461 | 3913470 | ||
BM544815.1 | - | 487 | ||||
CN365632.1 | - | 2 | 498 | |||
CV803721.1 | - | 527 |