Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRSK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032430.1 | NP_115806.1 | 16 | 2105 | ||
XM_005259327.3 | XP_005259384.1 | 16 | 2174 | |||
XM_011527395.2 | XP_011525697.2 | 14 | 1539 | |||
GenBank mRNA | AB058714.1 | - | 15 | 1640 | ||
AF479826.1 | - | 18 | 2003 | |||
AF479827.1 | - | 16 | 2105 | |||
AK124999.1 | - | 8 | 1074 | |||
AL831945.1 | - | 1 | 32 | |||
AL834275.1 | - | 16 | 1728 | |||
AY458602.1 | - | 16 | 1877 | |||
AY505124.1 | - | 16 | 1828 | |||
BC016681.1 | - | 8 | 1139 | 4341838 |