Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000574.4 | NP_000565.1 | 6 | 1069 | ||
NM_001114752.2 | NP_001108224.1 | 6 | 1069 | |||
NM_001300902.1 | NP_001287831.1 | 6 | 1069 | |||
NM_001300903.1 | NP_001287832.1 | 6 | 1069 | |||
NM_001300904.1 | NP_001287833.1 | 6 | 1069 | |||
NR_125349.1 | 6 | 1069 | ||||
XM_017000467.1 | XP_016855956.1 | 6 | 903 | |||
GenBank mRNA | AB240566.1 | - | 6 | 793 | ||
AB240567.1 | - | 6 | 793 | |||
AB240568.1 | - | 6 | 793 | |||
AB240569.1 | - | 6 | 808 | |||
AB240570.1 | - | 6 | 808 | |||
AF052110.1 | - | 6 | 1010 | |||
AK298549.1 | - | 7 | 771 | |||
AK300621.1 | - | 5 | 669 | |||
AK312448.1 | - | 6 | 864 | |||
AY055757.1 | - | 2 | 150 | |||
AY055758.1 | - | 4 | 390 | |||
AY055759.1 | - | 4 | 390 | |||
AY055760.1 | - | 4 | 390 | |||
BC001288.1 | - | 6 | 854 | 3460621 | ||
BT007159.1 | - | 6 | 775 | |||
M15799.1 | - | 6 | 760 | |||
M30142.1 | - | 6 | 840 | |||
M31516.1 | - | 6 | 840 | |||
U88576.1 | - | 5 | 673 |