Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142623.1 | NP_001136095.1 | 3, 4 | 498 | ||
NM_032484.4 | NP_115873.1 | 3, 4 | 498 | |||
GenBank mRNA | AF316997.1 | - | 1, 2 | 274 | ||
AK074700.1 | - | 2, 3 | 702 | |||
AK075277.1 | - | 3, 4 | 460 | |||
AK122777.1 | - | 4, 5 | 266 | |||
AK123075.1 | - | 659 | ||||
AK298933.1 | - | 3, 4 | 376 | |||
AK307628.1 | - | 2, 3 | 746 | |||
BC022784.1 | - | 3, 4 | 433 | 3847323 |