Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DCTN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199743.1 | NP_001186672.1 | 3 | 374 | ||
NM_032486.3 | NP_115875.1 | 3 | 374 | |||
NR_037573.1 | 4 | 475 | ||||
GenBank mRNA | AK027387.1 | - | 3 | 237 | ||
AK292843.1 | - | 3 | 301 | |||
AK299631.1 | - | 3 | 271 | |||
BC004191.2 | - | 3 | 254 | 3505855 | ||
CD388246.1 | - | 3 | 293 | |||
DA310703.1 | - | 3 | 374 | |||
DB087566.1 | - | 4 | 331 |