Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032526.2 | NP_115915.1 | 3 | 373 | ||
GenBank mRNA | AF331801.1 | - | 3 | 373 | ||
AY028778.1 | - | 3 | 364 | |||
BC103879.1 | - | 3 | 373 | 40004489 | ||
BC103880.1 | - | 3 | 373 | 40004491 |