Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: EBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278636.1 | NP_001265565.1 | 5 | 704 | ||
NM_032565.4 | NP_115954.1 | 4 | 642 | |||
NR_103802.1 | 5 | 763 | ||||
NR_103803.1 | 3 | 572 | ||||
GenBank mRNA | AF243433.1 | - | 4 | 631 | ||
BC018478.1 | - | 4 | 608 | 4476654 | ||
BC021021.1 | - | 5 | 681 | |||
BC073152.1 | - | 5 | 763 | |||
BC092471.1 | - | 4 | 637 | 4796715 | ||
BY799168.2 | - | 3 | 538 |