Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032649.5 | NP_116038.4 | 8 | 1171 | ||
GenBank mRNA | AJ417564.1 | - | 8 | 1004 | ||
AK098369.1 | - | 9 | 950 | |||
AK131263.1 | - | 2 | 819 | |||
AK289916.1 | - | 8 | 1026 | |||
AK303708.1 | - | 8 | 980 | |||
AK314572.1 | - | 8 | 1062 | |||
AY358756.1 | - | 8 | 1160 | |||
BC110295.1 | - | 8 | 1010 | 5196053 | ||
BC113512.1 | - | 8 | 999 | 8322564 | ||
BC117122.1 | - | 8 | 1002 | 40125673 |