Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC119B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080533.2 | NP_001074002.1 | 4 | 641 | ||
GenBank mRNA | AK126367.1 | - | 4 | 618 |