Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRINP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014618.2 | NP_055433.2 | 6 | 1172 | ||
GenBank mRNA | AF027734.1 | - | 6 | 1134 | ||
BC021560.1 | - | 6 | 1136 | |||
BC065196.1 | - | 6 | 1125 | 5245480 |