Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN4IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032730.5 | NP_116119.2 | 1 | 616 | ||
XM_017011375.1 | XP_016866864.1 | 1 | 616 | |||
XM_017011376.1 | XP_016866865.1 | 1 | 616 | |||
GenBank mRNA | AF336861.1 | - | 1 | 190 | ||
AF439711.1 | - | 1 | 139 | |||
AK313962.1 | - | 1 | 534 | |||
AY063761.1 | - | 1 | 139 | |||
BC006399.2 | - | 1 | 411 | 4309263 | ||
BQ936729.1 | - | 1 | 594 |