Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL17RC SIRNA Assays ›
Gene Symbol:
IL17RC
Gene Name:
interleukin 17 receptor C
Gene Aliases:
CANDF9, IL17-RL, IL17RL
Chromosome Location:
Chr.3: 9917074 - 9933627 on Build GRCh38
UniGene ID:
Hs.129959
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s39471
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IL17RC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001203263.1
NP_001190192.1
9
1020
NM_001203264.1
NP_001190193.1
9
1020
NM_001203265.1
NP_001190194.1
8
975
NM_032732.5
NP_116121.2
8
975
NM_153460.3
NP_703190.1
9
1020
NM_153461.3
NP_703191.1
9
1209
NR_037807.1
7
1186
XM_017007326.1
XP_016862815.1
9
1209
XM_017007327.1
XP_016862816.1
9
1209
XM_017007328.1
XP_016862817.1
9
1170
XM_017007329.1
XP_016862818.1
9
1209
XM_017007330.1
XP_016862819.1
9
1170
XM_017007331.1
XP_016862820.1
8
1164
XM_017007332.1
XP_016862821.1
8
1164
XM_017007333.1
XP_016862822.1
9
1209
XM_017007334.1
XP_016862823.1
8
1134
XM_017007335.1
XP_016862824.1
8
1125
XM_017007336.1
XP_016862825.1
9
1209
XM_017007337.1
XP_016862826.1
7
1089
XM_017007338.1
XP_016862827.1
8
1125
XM_017007339.1
XP_016862828.1
8
1164
XM_017007340.1
XP_016862829.1
9
997
XM_017007341.1
XP_016862830.1
8
952
XM_017007342.1
XP_016862831.1
8
921
XM_017007343.1
XP_016862832.1
8
921
XM_017007344.1
XP_016862833.1
9
958
XM_017007345.1
XP_016862834.1
8
913
XM_017007346.1
XP_016862835.1
8
952
XM_017007347.1
XP_016862836.1
7
876
XM_017007348.1
XP_016862837.1
8
913
XM_017007349.1
XP_016862838.1
7
876
GenBank mRNA
AF458065.1
-
9
975
AK075534.1
-
7
1151
AK092907.1
-
9
1210
AK096824.1
-
8
1117
AK225650.1
-
9
965
AK290670.1
-
8
1012
AK294616.1
-
6
1029
AK295377.1
-
7
994
AK303171.1
-
7
786
AK308241.1
-
8
905
AK315024.1
-
7
1163
AK315053.1
-
8
920
AY358840.1
-
8
937
AY359098.1
-
8
965
EF676032.1
-
8
886
EF676033.1
-
9
778
EF676034.1
-
9
789
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
cytokine-mediated signaling pathway
defense response to fungus
granulocyte chemotaxis
positive regulation of cytokine production involved in inflammatory response
positive regulation of interleukin-6 secretion
receptor binding
interleukin-17 receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0