Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032784.4 | NP_116173.2 | 2 | 452 | ||
XM_017011378.1 | XP_016866867.1 | 2 | 684 | |||
XM_017011379.1 | XP_016866868.1 | 2 | 605 | |||
GenBank mRNA | AF251057.1 | - | 2 | 444 | ||
AI095468.1 | - | 2 | 452 | |||
AK314912.1 | - | 2 | 223 | |||
BC022367.1 | - | 3 | 562 | 4733444 |