Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317895.1 | NP_001304824.1 | 4 | 1621 | ||
NM_032789.4 | NP_116178.2 | 5 | 1497 | |||
NR_134234.1 | 5 | 1497 | ||||
XM_011517336.2 | XP_011515638.1 | 5 | 1464 | |||
GenBank mRNA | AK027370.1 | - | 5 | 1454 | ||
AK075250.1 | - | 1 | 184 | |||
AK128537.1 | - | 6 | 1312 | |||
AK222914.1 | - | 5 | 1480 | |||
AK303317.1 | - | 4 | 1546 | |||
BC019030.2 | - | 1 | 317 | |||
BC020378.1 | - | 5 | 1472 | |||
BC030549.1 | - | 5 | 1471 | |||
BC132961.1 | - | 5 | 1448 | 40146983 | ||
BC136542.1 | - | 5 | 1448 | 9020532 | ||
BC144235.1 | - | 5 | 1448 |