Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ECI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178029.1 | NP_001171500.1 | 7 | 951 | ||
NM_001919.3 | NP_001910.2 | 7 | 1002 | |||
GenBank mRNA | AK291127.1 | - | 7 | 1002 | ||
BC000762.2 | - | 7 | 967 | 2821822 | ||
BC002746.2 | - | 7 | 922 | 3630566 | ||
BC009631.1 | - | 8 | 944 | 3877066 | ||
BC019316.1 | - | 7 | 967 | 2821822 | ||
BC020228.1 | - | 7 | 922 | 4541779 | ||
Z25820.1 | - | 7 | 975 |