Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032826.4 | NP_116215.1 | 2, 3 | 508 | ||
XM_011516645.2 | XP_011514947.1 | 2, 3 | 276 | |||
GenBank mRNA | AB052892.1 | - | 2, 3 | 484 | ||
AJ315497.1 | - | 2, 3 | 340 | |||
AJ315498.1 | - | 2, 3 | 330 | |||
AJ971941.1 | - | 2, 3 | 184 | |||
AK027603.1 | - | 2, 3 | 348 | |||
AK027726.1 | - | 1, 2 | 66 | |||
AK075368.1 | - | 2, 3 | 313 | |||
AK222874.1 | - | 2, 3 | 357 | |||
AK307621.1 | - | 2, 3 | 336 | |||
BC008413.1 | - | 2, 3 | 329 | 4333393 | ||
DA208815.1 | - | 2, 3 | 508 |