Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032836.2 | NP_116225.2 | 3 | 1935 | ||
XM_005259352.4 | XP_005259409.1 | 3 | 2960 | |||
XM_011527426.2 | XP_011525728.1 | 3 | 1896 | |||
GenBank mRNA | AK027674.1 | - | 3 | 1935 | ||
BC132777.1 | - | 2 | 1896 | 40146799 | ||
BC136512.1 | - | 2 | 1894 | 9020502 |