Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032859.2 | NP_116248.2 | 2 | 559 | ||
XM_011521128.2 | XP_011519430.1 | 2 | 517 | |||
GenBank mRNA | AK027812.1 | - | 1 | 16 | ||
AK124864.1 | - | 1 | 961 | |||
BC022566.1 | - | 2 | 475 | 4811611 | ||
BC070226.1 | - | 1 | 16 | 6716889 | ||
BQ424339.1 | - | 559 | ||||
BX647428.1 | - | 1 | 139 |