Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBASH3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032873.4 | NP_116262.2 | 2, 3 | 573 | ||
XM_005271712.3 | XP_005271769.1 | 2, 3 | 2819 | |||
GenBank mRNA | AB075839.1 | - | 2, 3 | 425 | ||
AK075203.1 | - | 2, 3 | 541 | |||
AK222843.1 | - | 2, 3 | 523 | |||
AK222846.1 | - | 2, 3 | 523 | |||
BC007541.1 | - | 2, 3 | 527 | 2958242 |