Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006933.1 | NP_001006934.1 | 3 | 969 | ||
NM_032926.2 | NP_116315.1 | 3 | 957 | |||
GenBank mRNA | AF163263.1 | - | 3 | 906 | ||
AK098494.1 | - | 3 | 900 | |||
BC007622.1 | - | 3 | 948 | 3355622 | ||
BC008703.1 | - | 3 | 957 | 2966921 |