Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297551.1 | NP_001284480.1 | 3 | 397 | ||
NM_001297552.1 | NP_001284481.1 | 3 | 397 | |||
NM_032927.3 | NP_116316.1 | 3 | 902 | |||
XM_005248034.2 | XP_005248091.1 | 3 | 420 | |||
GenBank mRNA | AK295248.1 | - | 3 | 379 | ||
AK296007.1 | - | 3 | 378 | |||
BC007729.2 | - | 3 | 867 | 4303698 | ||
BC091482.1 | - | 3 | 397 | 30344390 | ||
BM263799.1 | - | 3 | 379 | |||
CR933611.1 | - | 3 | 387 | |||
DA019506.1 | - | 3 | 387 |