Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SMIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032947.4 | NP_116565.3 | 2 | 1530 | ||
GenBank mRNA | AF313413.1 | - | 2 | 652 | ||
AK027847.1 | - | 2 | 717 | |||
AK125286.1 | - | 2 | 1530 | |||
BC009719.1 | - | 2 | 646 | |||
BC106040.1 | - | 2 | 1183 | 4940258 |