Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPIL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139126.3 | NP_624311.1 | 4, 5 | 418 | ||
GenBank mRNA | AF357880.1 | - | 4, 5 | 351 | ||
AK092069.1 | - | 4, 5 | 351 | |||
AK315388.1 | - | 4, 5 | 418 | |||
BC020986.1 | - | 4, 5 | 354 | 3852013 | ||
BC064134.1 | - | 4, 5 | 336 | |||
BC107684.1 | - | 4, 5 | 352 | |||
BX537536.1 | - | 4, 5 | 352 |