Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033402.4 | NP_208325.3 | 13 | 2287 | ||
XM_005251318.3 | XP_005251375.1 | 12 | 2076 | |||
XM_005251319.4 | XP_005251376.1 | 11 | 1770 | |||
XM_005251320.3 | XP_005251377.1 | 10 | 1562 | |||
XM_005251322.3 | XP_005251379.1 | 9 | 1376 | |||
XM_017013920.1 | XP_016869409.1 | 14 | 2235 | |||
XM_017013923.1 | XP_016869412.1 | 13 | 2062 | |||
XM_017013925.1 | XP_016869414.1 | 11 | 1781 | |||
XM_017013926.1 | XP_016869415.1 | 11 | 1764 | |||
XM_017013927.1 | XP_016869416.1 | 12 | 1792 | |||
XM_017013928.1 | XP_016869417.1 | 9 | 1244 | |||
GenBank mRNA | AB051551.1 | - | 12 | 2908 | ||
AK056185.1 | - | 12 | 2250 | |||
AK057990.1 | - | 4 | 542 | |||
AK300874.1 | - | 12 | 1931 | |||
AK302648.1 | - | 13 | 2137 | |||
AK307443.1 | - | 10 | 1543 | |||
AK307636.1 | - | 12 | 1921 | |||
BC030701.2 | - | 13 | 2160 | 4837403 |