Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TSPYL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033512.2 | NP_277047.2 | 1 | 221 | ||
GenBank mRNA | AB051537.1 | - | 1 | 160 | ||
AK091445.1 | - | 1 | 237 | |||
BC045630.1 | - | 1 | 221 | 5273220 |