Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031460.3 | NP_113648.2 | 4, 5 | 822 | ||
GenBank mRNA | AF339912.1 | - | 4, 5 | 788 | ||
AF358910.1 | - | 4, 5 | 682 | |||
AK315273.1 | - | 4, 5 | 813 | |||
AY358853.1 | - | 4, 5 | 773 | |||
BC025726.1 | - | 4, 5 | 769 | 5201326 | ||
EU978944.1 | - | 4, 5 | 729 |