Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHLDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134437.1 | NP_001127909.1 | 4 | 2022 | ||
NM_001134438.1 | NP_001127910.1 | 3 | 2080 | |||
NM_001134439.1 | NP_001127911.1 | 3 | 1841 | |||
NM_145753.2 | NP_665696.1 | 3 | 2080 | |||
GenBank mRNA | AB209903.1 | - | 3 | 1722 | ||
AF506820.1 | - | 2 | 1784 | |||
AJ496194.1 | - | 2 | 1695 | |||
AK092996.1 | - | 3 | 1766 | |||
AK096151.1 | - | 3 | 2022 | |||
AK125555.1 | - | 2 | 449 | |||
AK296841.1 | - | 4 | 2022 | |||
AK307491.1 | - | 4 | 1984 | |||
AK315050.1 | - | 3 | 1992 | |||
AL832205.1 | - | 3 | 1803 | |||
AL832476.1 | - | 3 | 1769 | |||
BC069194.1 | - | 3 | 1969 | |||
BC142678.1 | - | 2 | 1687 | 40148225 | ||
BC150210.1 | - | 2 | 1687 | 40148898 | ||
CR749654.1 | - | 4 | 2005 |