Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042632.2 | NP_001036097.1 | 4 | 2851 | ||
NM_001042633.2 | NP_001036098.1 | 5 | 2869 | |||
NM_033421.3 | NP_219489.1 | 4 | 2858 | |||
NM_152897.2 | NP_690857.1 | 5 | 1606 | |||
GenBank mRNA | AK095851.1 | - | 3 | 2729 |