Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042632.2 | NP_001036097.1 | 4 | 2366 | ||
NM_001042633.2 | NP_001036098.1 | 5 | 2384 | |||
NM_033421.3 | NP_219489.1 | 4 | 2373 | |||
NM_152897.2 | NP_690857.1 | 5 | 1121 | |||
XM_005260608.4 | XP_005260665.1 | 2 | 2448 | |||
XM_011529095.2 | XP_011527397.1 | 2 | 2352 | |||
GenBank mRNA | AK091359.1 | - | 3 | 2388 | ||
AK095851.1 | - | 3 | 2244 | |||
BC019823.1 | - | 5 | 1094 | 4453502 |