Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258304.1 | NP_001245233.1 | 5 | 974 | ||
NM_001258306.1 | NP_001245235.1 | 5 | 785 | |||
NM_138770.2 | NP_620125.1 | 5 | 983 | |||
XM_005263835.3 | XP_005263892.1 | 5 | 883 | |||
XM_005263836.4 | XP_005263893.1 | 3 | 1266 | |||
XM_005263837.1 | XP_005263894.1 | 5 | 901 | |||
XM_006712828.3 | XP_006712891.1 | 5 | 1082 | |||
XM_017005234.1 | XP_016860723.1 | 5 | 1095 | |||
XM_017005235.1 | XP_016860724.1 | 5 | 1154 | |||
XM_017005236.1 | XP_016860725.1 | 5 | 896 | |||
XM_017005237.1 | XP_016860726.1 | 5 | 989 | |||
XM_017005239.1 | XP_016860728.1 | 5 | 956 | |||
XM_017005240.1 | XP_016860729.1 | 5 | 1021 | |||
XM_017005241.1 | XP_016860730.1 | 6 | 891 | |||
XM_017005242.1 | XP_016860731.1 | 5 | 771 | |||
XM_017005244.1 | XP_016860733.1 | 6 | 687 | |||
XM_017005245.1 | XP_016860734.1 | 5 | 913 | |||
GenBank mRNA | AB299086.1 | - | 5 | 889 | ||
AK097664.1 | - | 2 | 2252 | |||
BC016861.1 | - | 5 | 898 | 3914851 | ||
BC030744.1 | - | 1 | 238 | |||
BC063387.1 | - | 5 | 722 | 5494806 |
Gene Symbol: CCDC74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258307.1 | NP_001245236.1 | 5 | 786 | ||
NM_207310.2 | NP_997193.1 | 5 | 984 | |||
XM_005263840.2 | XP_005263897.1 | 5 | 1009 | |||
XM_005263842.2 | XP_005263899.1 | 5 | 811 | |||
XM_006712833.2 | XP_006712896.1 | 5 | 1021 | |||
XM_006712834.3 | XP_006712897.1 | 5 | 961 | |||
XM_006712835.2 | XP_006712898.1 | 5 | 823 | |||
XM_006712837.2 | XP_006712900.1 | 5 | 763 | |||
XM_011512142.2 | XP_011510444.1 | 3 | 1252 | |||
GenBank mRNA | AK057609.1 | - | 5 | 908 | ||
AK297246.1 | - | 4 | 1896 | |||
AL133619.1 | - | 1 | 2820 | |||
BC067771.1 | - | 5 | 731 | 4824929 |