Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SPOCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281987.1 | NP_001268916.1 | 16 | 3459 | ||
NM_001281988.1 | NP_001268917.1 | 15 | 2037 | |||
NM_144569.5 | NP_653170.3 | 16 | 3498 | |||
XM_011542413.2 | XP_011540715.1 | 16 | 2028 | |||
XM_011542414.2 | XP_011540716.1 | 15 | 2223 | |||
XM_017002779.1 | XP_016858268.1 | 15 | 3365 | |||
XM_017002780.1 | XP_016858269.1 | 14 | 3326 | |||
XM_017002781.1 | XP_016858270.1 | 14 | 3242 | |||
XM_017002782.1 | XP_016858271.1 | 15 | 2597 | |||
GenBank mRNA | AK058077.1 | - | 6 | 1039 | ||
AK091016.1 | - | 15 | 2008 | |||
AK131447.1 | - | 16 | 3427 | |||
BC007910.2 | - | 8 | 1221 | |||
BC029069.1 | - | 6 | 1480 | |||
BC039870.2 | - | 1 | 263 | |||
BC114430.1 | - | 16 | 3385 | 40038407 |