Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPSB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324081.1 | NP_001311010.1 | 7 | 1480 | ||
NM_080861.3 | NP_543137.2 | 7 | 1502 | |||
XM_005255673.4 | XP_005255730.1 | 7 | 1692 | |||
GenBank mRNA | AK057850.1 | - | 1527 | |||
AK074190.1 | - | 7 | 1466 | |||
BC007588.1 | - | 7 | 1448 | 3342825 | ||
BC024244.1 | - | 7 | 1466 | 4798873 | ||
BC035065.1 | - | 7 | 1459 | |||
BC041897.1 | - | 6 | 2276 | 5298729 | ||
BC047441.1 | - | 7 | 1444 | 5303706 | ||
BC065515.1 | - | 4 | 1745 | |||
BQ930544.1 | - | 2 | 783 |