Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIFC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145754.3 | NP_665697.1 | 17 | 2648 | ||
XM_005272357.3 | XP_005272414.1 | 18 | 2658 | |||
GenBank mRNA | AK055045.1 | - | 13 | 1741 | ||
AY007121.1 | - | 1 | 910 | |||
BC015171.2 | - | 1 | 925 | |||
BC017311.2 | - | 18 | 2630 | 5090622 |